missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ Malt Extract
GREENER_CHOICE

Catalog No. 218610TF
Change view
Click to view available options
Quantity:
500 g
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
218610TF 10 kg
DF0186177 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 218610TF Supplier Gibco™ Supplier No. 218610

The water-soluble portion of malted barley.

  • Bacto Malt Extract is the water-soluble portion of malted barley
  • Due to the extraction process, this animal origin-free supplement provides a very high carbohydrate content to any medium
  • It has been used successfully in microbial fermentation applications
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C

Specifications

Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Media Supplement
Product Line Bacto
Quantity 10 kg
Source Animal origin-free, malted barley