missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ Casitone
GREENER_CHOICE

Catalog No. DF0259179
Change view
Click to view available options
Quantity:
500 g
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0259179 500 g
225910 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. DF0259179 Supplier Gibco™ Supplier No. 225930

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An animal origin (AO) pancreatic enzymatic digest of casein.

  • An animal origin (AO) pancreatic enzymatic digest of casein similar to Difco Casein Digest
  • This supplement provides a nutritional profile based on casein
  • The specific enzymatic digest performed for this product results in a particularly high content of amino acids and peptides of varying sizes, making it a more nutritious hydrolysate
  • It has been successfully used for the production of Escherichia coli-based recombinant proteins
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Dehydrated Culture Media
Format Bottle
For Use With (Application) Production of Escherichia coli-based recombinant proteins
Product Line Bacto
Quantity 500 g
Source Animal origin, casein
What alternative product do you recommend for Bacto Tryptone (Cat. No. 211699, 211701, 211705)?

We have several casein digests that could serve as alternatives to Bactone Tryptone (Cat. No. 211699, 211791, 211705), including Bacto Casitone (Cat. No. 225930, 225910), Difco Tryptone Peptone (Cat. No. 21921, 211922, 211923), BiTek Tryptone (Cat. No. 251420). However, the digestion and nutritive profile of each peptone are different, therefore there may be differences in bacterial growth or supplementation ability. We recommend empirically determining the performance of alternative casein digestion in your application.