missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bacteriophage M13 Monoclonal Antibody (K70008_8E2), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA556370
Description
Tail virion protein G7P (Bacteriophage M13) may initiate with G9P the virion concomitant assembly-budding process, by interacting with the packaging signal of the viral genome. The assembly-budding takes place at the host inner membrane. In turn, G7P and G9P are present at the end of the filamentous virion that emerges first from the bacterial host. Anti-Bacteriophage M13 Antibody is useful for researchers interested in HCP, Host Cell Membranes, Transmembrane Viruses, and E. coli bacteria.Chemical Identifiers
| Bacteriophage M13 | |
| 0.77 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| M13 bacteriophage. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| K70008_8E2 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Virus | |
| Enterobacteria phage M13 | |
| IgG2a | |
| Protein A | |
| Antibody |
Specifications
| Bacteriophage M13 | |
| Monoclonal | |
| 0.77 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2a |
| Western Blot | |
| K70008_8E2 | |
| Unconjugated | |
| Enterobacteria phage M13 | |
| M13 bacteriophage. | |
| 200 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |