missing translation for 'onlineSavingsMsg'
Learn More

Baclofen 98.0+%, TCI America™

Catalog No. p-7122888
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33435G 5 g
B334325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33435G Supplier TCI America Supplier No. B33435G

Chemical Identifiers

CAS 1134-47-0
Molecular Formula C10H12ClNO2
Molecular Weight (g/mol) 213.661
MDL Number MFCD00055143
InChI Key KPYSYYIEGFHWSV-UHFFFAOYSA-N
Synonym baclofen, lioresal, baclon, 4-amino-3-4-chlorophenyl butanoic acid, dl-baclofen, kemstro, gabalon, baclofene, baclofeno, baclofenum
PubChem CID 2284
ChEBI CHEBI:2972
IUPAC Name 4-amino-3-(4-chlorophenyl)butanoic acid
SMILES C1=CC(=CC=C1C(CC(=O)O)CN)Cl

Specifications

Melting Point 208°C
UN Number 2811
Quantity 5 g
Formula Weight 213.66
Percent Purity ≥98.0% (T)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material Baclofen
RTECSNumber MW5084200
TSCA No