Learn More
Mopec Back Draft Countertop Dissection Work Bench Grossing Station
Available in four highly customizable models, these back draft vented workstations help control odors, gases and formalin fumes. Mopec™ Back Draft Countertop Dissection Work Bench Grossing Station ensures a safe laboratory working environment.
Supplier: Mopec HL048
Description
- Countertop constructed of 14 gauge stainless steel with a recessed edge
- Includes vent tie-in
- Adjustable leveling feet
- GFCI outlet
Warranty
1 Year Warranty
Spécifications
| Ventilated Grossing Station | |
| 30 in. | |
| 62 in. | |
| 157.48 cm |
| Stainless Steel (14 ga.) | |
| 76.2 cm | |
| 48 in. | |
| 121.92 cm |