missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BAAT1 Recombinant Rabbit Monoclonal Antibody (23GB4355)
GREENER_CHOICE

Catalog No. MA552696
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552696 100 μL
MA552697 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552696 Supplier Invitrogen™ Supplier No. MA552696
Only null left

Rabbit Recombinant Monoclonal Antibody

BAAT1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Required for activation of ATM following ionizing radiation. May act by regulating dephosphorylation of ATM.

Specifications

Antigen BAAT1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4355
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BRAT1
Gene Accession No. Q6PJG6
Gene Alias AA881470; BAAT1; Brat1; Brat1-ps1; BRCA1 associated ATM activator 1; BRCA1-associated ATM activator 1; BRCA1-associated ATM activator 1, pseudogene 1; BRCA1-associated protein required for ATM activation protein 1; C7orf27; HEAT repeat-containing protein C7orf27; MGC22916; RGD1565146; RMFSL
Gene Symbols BRAT1
Host Species Rabbit
Immunogen A synthesized peptide derived from human BRAT1 (701-821AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221927
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less