Learn More
Motic™ BA310E Compound LED Microscopes
Catalog No. 01258024
Change view
Click to view available options
Head Style:
Binocular
Trinocular
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Head Style
01258024 Trinocular
01258023 Binocular
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01258024 Supplier Motic™ Supplier No. 1100100402641
Committed to the concept of continuously improving its models, Motic™’s Elite series microscopes feature the Infinity Optical System and improved CCIS™ EC Plan Objective Lenses. Provides superior image clarity and contrast.
These models introduce a new rackless stage concept without a prominent gear rack to allow for a more convenient use of the X/Y movement.
  • Allow easy interchangeability of its halogen bulb with LED modules
  • Swiveling, 30° inclined Siedentopf type
  • Reversed quintuple revolving nosepiece
  • Coaxial coarse and fine focusing adjustment with tension adjustment
  • Built in low position coaxial rackless stage (Right-hand control)
  • Diopter Adjustment: ±5
Specifications
Voltage 6 V
Magnification Power 10X
Objective Types CCIS™ EC Plan Achromat Objectives EC PL (4X, 10X, 40X(S), 100X(S-oil))
Nosepiece Reversed Quadruple
Head Style Trinocular
Lighting Type LED
Eyepiece WF 10X/20 mm