Learn More
Motic™ BA310 Binocular Digital Microscope
Description
- Built-in, live digital imaging camera with high resolution
- USB 2.0
- Revolving nosepiece
- Coaxial coarse and fine focusing adjustment with tension adjustment
- Built-in, low position coaxial mechanical stage (right-hand control)
- Focusable Abbe condenser with 0.9 numerical aperature
- Iris diaphragm and slot
- Diopter Adjustment: ±5
Specifications
Specifications
| Resolution | 5 MP |
| Magnification Power | 10X |
| Objective Types | CCIS™ EC Plan Achromat Objectives (4X, 10X, 40X(S), 100X(S-oil)) |
| Nosepiece | Reversed Quintuple |
| Head Style | Binocular |
| Lighting Type | Kohler Illumination |
| Eyepiece | WF 10X/20 mm |