Learn More
Motic BA210S LED Compound Microscope
The BA210S biological compound microscope is designed for advanced biology students, teachers, doctors, and lab technicians.
Supplier:  Motic 1100100403841
 View more versions of this product
Catalog No. 01257473
Only null left
Description
Description
  • Every aspect of these microscopes contributes to making your daily work easier.
  • The ergonomic shape of the stand and the comfortability of the different heads enables fatigue-free viewing for many hours.
Specifications
Chemical Identifiers
10X
4X/0.10 (DT 17mm), 10X/0.25 (DT 6.4mm), 40X/0.65/S (DT 0.45mm), 100X/1.25/S (DT 0.14mm)
LED
WF 10X/20 mm
Specifications
10X
Trinocular
WF 10X/20 mm
4X/0.10 (DT 17mm), 10X/0.25 (DT 6.4mm), 40X/0.65/S (DT 0.45mm), 100X/1.25/S (DT 0.14mm)
LED
Product Suggestions
Product Suggestions