Learn More
Brady™ B33 Series Glossy White Component and Barcode Labels
Permanent polyester (B-423) is ideal for electronic PCB and component identification, barcode labels, rating plates and solar panel identification.
$171.80 - $383.37
Specifications
| For Use With (Equipment) | Brady Label Printers |
|---|---|
| Material | Polyester (B-423) |
| Print Technology | Thermal Transfer |
| Thickness (English) | 0.003 in. |
| Adhesive Type | Permanent Acrylic |
| Catalog Number | Mfr. No. | Height (English) | Labels per Row | Width (English) | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Height (English) | Labels per Row | Width (English) | Price | Quantity & Availability | |||
|
17000975
|
Brady™
B3370423 |
0.25 in. | 7 | 0.25 in. |
Each for $183.15
|
|
|||
|
17000970
|
Brady™
B332423 |
0.25 in. | 3 | 0.9 in. |
Each for $245.65
|
|
|||
|
17000974
|
Brady™
B3351423 |
0.25 in. | 3 | 1 in. |
Each for $245.65
|
|
|||
|
17000972
|
Brady™
B3313423 |
0.25 in. | 2 | 1.25 in. |
Each for $194.52
|
|
|||
|
17000971
|
Brady™
B334423 |
0.25 in. | 2 | 1.5 in. |
Each for $239.94
|
|
|||
|
17000973
|
Brady™
B3315423 |
0.25 in. | 1 | 2 in. |
Each for $171.80
|
|
|||
|
17001006
|
Brady™
B33340423 |
0.375 in. | 7 | 0.375 in. |
Each for $188.85
|
|
|||
|
17000976
|
Brady™
B33147423 |
0.375 in. | 2 | 1.5 in. |
Each for $230.02
|
|
|||
|
17000977
|
Brady™
B3312423 |
0.437 in. | 5 | 0.5 in. |
Each for $238.56
|
|
|||
|
17000978
|
Brady™
B3337423 |
0.5 in. | 2 | 1.5 in. |
Each for $255.57
|
|
|||
|
17000979
|
Brady™
B336423 |
0.75 in. | 2 | 1.5 in. |
Each for $232.85
|
|
|||
|
17000980
|
Brady™
B337423 |
1.25 in. | 1 | 2.75 in. |
Each for $188.85
|
|
|||
|
17000981
|
Brady™
B3376423 |
3 in. | 1 | 4 in. |
Each for $336.52
|
|
|||
|
17000982
|
Brady™
B33161423 |
4 in. | 1 | 4 in. |
Each for $362.07
|
|
|||
|
17000983
|
Brady™
B3320423 |
5 in. | 1 | 3 in. |
Each for $275.44
|
|
|||
|
17000984
|
Brady™
B3325423 |
6 in. | 1 | 4 in. |
Each for $383.37
|
|
|||
Description
Permanent polyester (B-423) is ideal for electronic PCB and component identification, barcode labels, rating plates and solar panel identification. A ribbon is required to print this material.
- Glossy white finish with permanent acrylic is thermal transfer printable
- UL approved, CSA accepted
- Meets requirements of a halogen-free material per DIN VDE 0472 part 815
- Wide service temperature range
Specifications
| Brady Label Printers | |
| Thermal Transfer | |
| Permanent Acrylic | |
| Non-sterile | |
| Die-cut | |
| -94°F to 230°F |
| Polyester (B-423) | |
| 0.003 in. | |
| Autoclavable | |
| Gloss | |
| Single-side Printable | |
| Barcode Labeling, Equipment Labeling, Data and Telecommunications Labeling, Inventory and Inspection Labeling, General Identification |