Learn More
United Scientific™ B.O.D. Bottles, Unnumbered
Description
- Pack of 36 unnumbered glass BOD bottles with 60 mL capacity.
- Ideal for incubating diluted samples of clean, waste, effluent and polluted water during the five-day biochemical oxygen demand (BOD) test.
- Glass pennyhead stoppers included.
Specifications
Specifications
| Volume (Metric) | 60 mL |
| Stopper | Glass Pennyhead |