Learn More
United Scientific™ B.O.D. Bottles, Unnumbered
BOD Bottles for incubating diluted samples of clean, waste, effluent, and polluted water during the 5-day biochemical oxygen demand (BOD) test.
Supplier:  United Scientific™ BOD300
 View more versions of this product
Catalog No. S15753
Only null left
Description
Description
  • Pack of 36 unnumbered glass BOD bottles with 60 mL capacity.
  • Ideal for incubating diluted samples of clean, waste, effluent and polluted water during the five-day biochemical oxygen demand (BOD) test.
  • Glass pennyhead stoppers included.
Specifications
Chemical Identifiers
Glass Pennyhead
Specifications
300 mL
Glass Pennyhead
Product Suggestions
Product Suggestions