missing translation for 'onlineSavingsMsg'
Learn More

(+)-B-Chlorodiisopinocampheylborane (58% in Hexane, ca. 1.6mol/L), TCI America™

Catalog No. C1614100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1614100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C1614100ML Supplier TCI America Supplier No. C1614100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112246-73-8
Molecular Formula C20H34BCl
Molecular Weight (g/mol) 320.75
MDL Number MFCD00074808
InChI Key PSEHHVRCDVOTID-NAVXHOJHSA-N
Synonym +-b-chlorodiisopinocamphenylborane
PubChem CID 133124869
IUPAC Name chlorobis[(1S,2R,3S,5S)-2,6,6-trimethylbicyclo[3.1.1]heptan-3-yl]borane
SMILES C[C@H]1[C@@H]2C[C@H](C[C@@H]1B(Cl)[C@H]1C[C@H]3C[C@@H]([C@@H]1C)C3(C)C)C2(C)C

Specifications

Color Yellow
Quantity 100 mL
UN Number 2924
Formula Weight 320.75
Physical Form Liquid
Chemical Name or Material (+)-B-Chlorodiisopinocampheylborane (58% in Hexane, ca. 1.6mol/L)
TSCA No
Recommended Storage Refrigerator