missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B-Bromocatecholborane 97.0+%, TCI America™
Supplier: TCI America B213225G
Specifications
| B-Bromocatecholborane | |
| 52°C | |
| 76°C | |
| MFCD00013341 | |
| 2925 | |
| SAILTPCYIYNOEL-UHFFFAOYSA-N | |
| 2-bromo-1,3,2-benzodioxaborole | |
| 3552384 | |
| ≥97.0% (T) |
| 51901-85-0 | |
| Purple-White | |
| C6H4BBrO2 | |
| 25 g | |
| 2-Bromo-1,3,2-benzodioxaborole | |
| B1(OC2=CC=CC=C2O1)Br | |
| 198.81 | |
| 198.81 | |
| Crystalline Powder |
Chemical Identifiers
| 51901-85-0 | |
| 198.81 | |
| SAILTPCYIYNOEL-UHFFFAOYSA-N | |
| 3552384 | |
| B1(OC2=CC=CC=C2O1)Br |
| C6H4BBrO2 | |
| MFCD00013341 | |
| 2-Bromo-1,3,2-benzodioxaborole | |
| 2-bromo-1,3,2-benzodioxaborole |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator