accessibility menu, dialog, popup

UserName

BOOBr

CAS RN 51901-85-0

IUPAC Name: 2-bromo-2H-1,3,2-benzodioxaborole
Synonyms: 2-bromo-1,3,2-benzodioxaborole
Molecular Weight (g/mol): 198.81
Molecular Formula: C6H4BBrO2
InChi Key: SAILTPCYIYNOEL-UHFFFAOYSA-N
SMILES: BrB1OC2=CC=CC=C2O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results