missing translation for 'onlineSavingsMsg'
Learn More

B-Bromocatecholborane 97.0+%, TCI America™

Catalog No. B21325G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21325G 5 g
B213225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21325G Supplier TCI America Supplier No. B21325G

Chemical Identifiers

CAS 51901-85-0
Molecular Formula C6H4BBrO2
Molecular Weight (g/mol) 198.81
MDL Number MFCD00013341
InChI Key SAILTPCYIYNOEL-UHFFFAOYSA-N
Synonym 2-Bromo-1,3,2-benzodioxaborole
PubChem CID 3552384
IUPAC Name 2-bromo-1,3,2-benzodioxaborole
SMILES B1(OC2=CC=CC=C2O1)Br

Specifications

Melting Point 52°C
Color Purple-White
Boiling Point 76°C
Quantity 5 g
UN Number 2925
Formula Weight 198.81
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material B-Bromocatecholborane
TSCA No
Recommended Storage Refrigerator