missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ B-27™ Supplement, minus insulin

Description
B-27 Supplement is suitable for use in most neuronal cell applications. For other applications, we provide alternative formulations of B-27 Supplement and Neurobasal media.
For established protocols using B-27 Supplement, access our Gibco Neurobiology Protocol Handbook.
View publications using Gibco neurobiology products at our Gibco CellCite.
Specifications
Specifications
| Content And Storage | Store in freezer (-5 to -20°C) and protect from light. |
| Cell Type | Neural Cells |
| Classification | Serum-free |
| Concentration | 50 X |
| Culture Type | Mammalian Cell Culture |
| Form | Liquid |
| Product Type | Cell Culture Supplement |
| Serum Level | Serum-Free |
| Sterility | Sterile |
| Product Line | B-27 |
| Show More |
Frequently Asked Questions (FAQs)
Do you offer B-27 supplements with and without insulin?
Yes, we do offer B-27 Supplement, minus insulin (Cat. Nos. A1895601 and A1895602). All our other B-27 supplements do contain insulin.
For Research Use Only. Not for use in diagnostic procedures.