missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ B-27™ Supplement (50X), minus vitamin A

Catalog No. p-7075643
Encompass
Change view
Click to view available options
Quantity:
10 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12587010 10 mL
12587001 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12587010 Supplier Gibco™ Supplier No. 12587010
Only null left

A variant of the original serum-free B-27 Supplement, designed for the proliferation of neural stem cells in the absence of vitamin A.

Gibco B-27 Supplement minus vitamin A is a customized, serum-free formulation based on the original, most cited neuronal cell culture supplement, designed specifically to support the proliferation and cultivation of neural stem cells (NSCs) and neural progenitor cells. By excluding vitamin A (also known as retinyl acetate or retinol), this variant prevents the differentiation-inducing effects of retinoic acid, making it ideal for maintaining NSCs and neural progenitors in their undifferentiated state.

  • Optimized for NSC proliferation—by removing vitamin A, this supplement is perfectly suited for the growth and expansion of embryonic and adult neural stem cells and neural precursors, without inducing differentiation
  • Versatile cultivation—supports the cultivation of neural progenitor cells and NSCs in both neurospheres in suspension and adherent monolayer cultures, providing flexibility in your experimental design. Ideal for cell culture where retinoic acid signaling must be controlled.
  • High quality—cGMP-manufactured to ensure the highest standards of quality and consistency. Each batch undergoes rigorous performance testing, including assessments of its ability to support the differentiation of human neural stem cells to neurons.
  • Comprehensive testing—every lot is tested for endotoxins, sterility, and mycoplasma to ensure the utmost safety and reliability
  • Proven performance—derived from the proprietary formulation of the original B-27 Supplement, trusted by researchers worldwide for its consistent and reliable results

Choose Gibco B-27 Supplement, minus vitamin A, for an optimized, high-quality solution that supports the proliferation of neural stem cells while preventing unwanted differentiation influenced by retinoic acid, ensuring consistent and reliable results for your research.

Specifications

Content And Storage Store in freezer (-5 to -20°C) and protect from light.
Cell Type Neural Progenitor Cells
Classification Serum-free
Concentration 50 X
Culture Type Mammalian Cell Culture
Form Liquid
Product Type Cell Culture Supplement
Serum Level Serum-Free
Sterility Sterile
Product Line B-27
Quantity 10 mL
Shipping Condition Dry Ice
Source Serum-Free
Tested For Endotoxin, Performance, Sterility, Mycoplasma
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.