missing translation for 'onlineSavingsMsg'
Learn More

Azure Biosystems AzureSpectra Labeling Kit, for Western Blot

Catalog No. AC2187AZ
Change view
Click to view available options
Label or Dye:
490 Dye
550 Dye
650 Dye
700 Dye
800 Dye
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Label or Dye
AC2187AZ 650 Dye
AC2186AZ 550 Dye
AC2188AZ 700 Dye
AC2189AZ 800 Dye
AC2185AZ 490 Dye
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC2187AZ Supplier Azure Biosystems Supplier No. AC2187
Only null left

Kit for covalently labeling antibodies for Western blot with a fluorescent dye. Contains labeling buffer, dye solution, quenching solution, neutralization buffer and two G-25 columns. Each kit includes sufficient reagents to perform two labelings of 50 micrograms of antibody

  • For fast and simple antibody labeling
  • AzureSpectra Labeling kits provide all the materials needed to quickly label your primary and secondary antibodies with fluorescent dyes. Choose between AzureSpectra dyes 490, 550, 650, 700 or 800.
  • AzureSpectra Labeling Kits include enough reagents to label 100 μg of antibody (2 reactions of 50 μg each):
    • Antibody Labeling Buffer
    • Dye Solution
    • Quenching Solution
    • Neutralization Buffer
    • 2 G-25 Spin Columns

Specifications

Detection Method Fluorescence
Product Type Protein Labeling Reagent
Membrane Compatibility Any
Label or Dye 650 Dye