Learn More
Azure Biosystems AzureSpectra Labeling Kit, for Western Blot
Kit for covalently labeling antibodies for Western blot with a fluorescent dye. Contains labeling buffer, dye solution, quenching solution, neutralization buffer and two G-25 columns. Each kit includes sufficient reagents to perform two labelings of 50 micrograms of antibody
Supplier: Azure Biosystems AC2186
Description
- For fast and simple antibody labeling
- AzureSpectra Labeling kits provide all the materials needed to quickly label your primary and secondary antibodies with fluorescent dyes. Choose between AzureSpectra dyes 490, 550, 650, 700 or 800.
- AzureSpectra Labeling Kits include enough reagents to label 100 μg of antibody (2 reactions of 50 μg each):
- Antibody Labeling Buffer
- Dye Solution
- Quenching Solution
- Neutralization Buffer
- 2 G-25 Spin Columns
Chemical Identifiers
| Fluorescence | |
| Any | |
| Protein Labeling Reagent |
Specifications
| Fluorescence | |
| Protein Labeling Reagent | |
| 550 Dye | |
| Any |