missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azure Biosystems AzureSpectra Fluorescent Western Blot Kit, with Fluorescent Block
Kit contains materials for 10 fluorescent Western blots, including LF PVDF membranes, wash buffer, fluorescent blot blocking buffer, and secondary antibodies.
$1145.97 - $1145.97
Specifications
| Product Type | Western Blot Kit |
|---|---|
| Detection Method | Fluorescence |
| For Use With (Application) | Western Blot |
| Membrane Compatibility | Nitrocellulose, PVDF |
| Quantity | 10 Mini Blots |
Description
- Full kits for two-color fluorescent Western blots.
- Also offered with Fluorescent Block.
Specifications
| Western Blot Kit | |
| Western Blot | |
| 10 Mini Blots |
| Fluorescence | |
| Nitrocellulose, PVDF |