missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azure Biosystems AzureSpectra Fluorescent Western Blot Kit, with Fluorescent Block
Description
- Full kits for two-color fluorescent Western blots.
- Also offered with Fluorescent Block.
Specifications
Specifications
| Product Type | Western Blot Kit |
| Detection Method | Fluorescence |
| For Use With (Application) | Western Blot |
| Membrane Compatibility | Nitrocellulose, PVDF |
| Quantity | 10 Mini Blots |
| Kit Contents | Includes: Secondary Ab rb650/ms550 |
| Content And Storage | LF PVDF Membranes, Wash Buffer, Fluorescent Blot Blocking Buffer, Secondary Antibodies rb650/ms550 |