missing translation for 'onlineSavingsMsg'
Learn More

Azure II, TCI America™

Catalog No. A05741G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A05741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A05741G Supplier TCI America Supplier No. A05741G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 37247-10-2
MDL Number MFCD00080716
Synonym Methylene Azure II

Specifications

Melting Point 185°C
Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material Azure II