accessibility menu, dialog, popup

UserName

H3CNCH3NSNCH3CH3

CAS RN 37247-10-2

IUPAC Name: 3,7-bis(diethylamino)-5λ⁴-phenothiazin-5-ylium
Synonyms: 3,7-bis(diethylamino)-5λ⁴-phenothiazin-5-ylium
Molecular Weight (g/mol): 340.51
Molecular Formula: C20H26N3S
InChi Key: JXBCJUPBVRELNY-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(CC)CC
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results