missing translation for 'onlineSavingsMsg'
Learn More

Azulene, MilliporeSigma™ Supelco™

Catalog No. 111020394
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020394 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020394 Supplier Merck Emd Millipore Supplier No. 3787950MG
Environmental

Specifications

CAS 275-51-4
Physical Form Neat
Linear Formula C10H8
MDL Number MFCD00003810
Melting Point 98°C to 100°C
Boiling Point 242°C
Synonym Bicyclo[5.3.0 ]decapentaene
Type Analytical Standard
Quantity 50 mg
Formula Weight 128.17
Grade Analytical Standard
Show More Show Less