missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Azoxymethane

Catalog No. 0218397180
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
0218397180 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0218397180 Supplier MP Biomedicals Supplier No. 0218397180

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Induces colon tumorigenesis in lab animals

  • A carcinogen that induces O6-methylguanine adducts in DNA leading to G→A transitions
  • Induces tumorigenesis in the colon of laboratory animals
  • Used to study the mechanism or cancer progression and chemoprevention
  • For research or further manufacturing use only, not for direct human use

Specifications

Content And Storage -20°C
Classification GHS08
Form Liquid
Product Type Carcinogen
Sterility Non-sterile
For Use With (Application) Induces tumorigenesis in the colon of laboratory animals and is used to study the mechanism of cancer progression and chemoprevention
Quantity 100 mg
Regulatory Status Research Use or Further Manufacturing

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.