Learn More
Azomethine-H hydrate
Reagent for boron determination | CAS: 32266-60-7 | C17H13NO8S2 | 423.41 g/mol
$111.26 - $826.96
Chemical Identifiers
| CAS | 32266-60-7 |
|---|---|
| Molecular Formula | C17H13NO8S2 |
| Molecular Weight (g/mol) | 423.41 |
| MDL Number | MFCD00066536 |
| InChI Key | DUCCKQSNXPFEGT-NVMNQCDNSA-N |
| PubChem CID | 5702781 |
| SMILES | OC1=CC=CC=C1\C=N/C1=C2C(O)=CC(=CC2=CC(=C1)S(O)(=O)=O)S(O)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA1681004
|
Thermo Scientific Chemicals
01681004 |
2 g |
Each for $111.26
|
|
|||||
|
AA1681009
|
Thermo Scientific Chemicals
01681009 |
10 g |
Each for $393.45
|
|
|||||
|
AA1681014
|
Thermo Scientific Chemicals
01681014 |
25 g |
Each for $826.96
|
|
|||||
Description
Azomethine-H hydrate is used as a highly sensitive and colorimetric reagent for testing and detection of boron. It is also used to detect microgram levels of boron in glass and steel samples. Further, it is involved in the detection of boron in plant samples by masking copper, iron and aluminum ions with ethylenediaminetetraacetic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 32266-60-7 | |
| 423.41 | |
| DUCCKQSNXPFEGT-NVMNQCDNSA-N | |
| OC1=CC=CC=C1\C=N/C1=C2C(O)=CC(=CC2=CC(=C1)S(O)(=O)=O)S(O)(=O)=O |
| C17H13NO8S2 | |
| MFCD00066536 | |
| 5702781 |
Specifications
| 32266-60-7 | |
| MFCD00066536 | |
| DUCCKQSNXPFEGT-NVMNQCDNSA-N | |
| 4-hydroxy-5-[[(Z)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]naphthalene-2,7-disulfonic acid | |
| 5702781 | |
| Light sensitive | |
| Powder |
| C17H13NO8S2 | |
| Soluble in acetone,ammonium hydroxide,n-butanol and water. | |
| OC1=CC=CC=C1\C=N/C1=C2C(O)=CC(=CC2=CC(=C1)S(O)(=O)=O)S(O)(=O)=O | |
| 423.41 | |
| 423.40 (Anhydrous) | |
| 2 g | |
| Azomethine-H hydrate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 250-975-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only