accessibility menu, dialog, popup

UserName

OHNOHSOHOOSHOOO(Z)

CAS RN 32266-60-7

IUPAC Name: 4-hydroxy-5-[(Z)-[(2-hydroxyphenyl)methylidene]amino]naphthalene-2,7-disulfonic acid
Synonyms: 4-hydroxy-5-[(Z)-[(2-hydroxyphenyl)methylidene]amino]naphthalene-2,7-disulfonic acid
Molecular Weight (g/mol): 423.41
Molecular Formula: C17H13NO8S2
InChi Key: DUCCKQSNXPFEGT-NVMNQCDNSA-N
SMILES: OC1=CC=CC=C1\C=N/C1=C2C(O)=CC(=CC2=CC(=C1)S(O)(=O)=O)S(O)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)