missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azoic Diazo Component 24 (Salt) 95.0+%, TCI America™
Supplier: TCI America B07855G
Specifications
| Azoic Diazo Component 24 (Salt) [for Biochemical Research] | |
| Green-Yellow | |
| MFCD00074762 | |
| 4-Benzamido-2,5-dimethoxybenzenediazonium Salt Zinc Chloride, Fast Blue RR Salt | |
| COC1=CC(=C(C=C1N=C(C2=CC=CC=C2)[O-])OC)[N+]#N.COC1=CC(=C(C=C1N=C(C2=CC=CC=C2)[O-])OC)[N+]#N.Cl.Cl.[Cl-].[Cl-].[Zn+2] | |
| 775.77 | |
| 775.77 | |
| Crystalline Powder |
| 55663-99-5 | |
| C30H28Cl4N6O6Zn | |
| 5 g | |
| LLARVKAOQLUCQI-UHFFFAOYSA-L | |
| zinc;N-(4-diazonio-2,5-dimethoxyphenyl)benzenecarboximidate;dichloride;dihydrochloride | |
| 134159107 | |
| ≥95.0% (T) |
Chemical Identifiers
| 55663-99-5 | |
| 775.77 | |
| LLARVKAOQLUCQI-UHFFFAOYSA-L | |
| 134159107 | |
| COC1=CC(=C(C=C1N=C(C2=CC=CC=C2)[O-])OC)[N+]#N.COC1=CC(=C(C=C1N=C(C2=CC=CC=C2)[O-])OC)[N+]#N.Cl.Cl.[Cl-].[Cl-].[Zn+2] |
| C30H28Cl4N6O6Zn | |
| MFCD00074762 | |
| 4-Benzamido-2,5-dimethoxybenzenediazonium Salt Zinc Chloride, Fast Blue RR Salt | |
| zinc;N-(4-diazonio-2,5-dimethoxyphenyl)benzenecarboximidate;dichloride;dihydrochloride |
Safety and Handling
EINECSNumber : (5)-2314
TSCA : No