missing translation for 'onlineSavingsMsg'
Learn More

Azobenzene 98.0+%, TCI America™

Supplier:  TCI America A0565100G

 View more versions of this product

Catalog No. A0565100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Azobenzene
68°C
C12H10N2
100 g
azobenzene, diazene, diphenyl, diazobenzene, e-diphenyldiazene, azobisbenzene, azofume, 1,2-diphenyldiazene, azodibenzeneazofume, diphenyldiimide, azobenzide
C1=CC=C(C=C1)N=NC1=CC=CC=C1
182.23
CHEBI:190358
≥98.0% (GC)
103-33-3
293°C
MFCD00003022
3077
DMLAVOWQYNRWNQ-UHFFFAOYSA-N
diphenyldiazene
2272
182.23
Crystalline Powder

Chemical Identifiers

103-33-3
182.23
DMLAVOWQYNRWNQ-UHFFFAOYSA-N
2272
diphenyldiazene
C12H10N2
MFCD00003022
azobenzene, diazene, diphenyl, diazobenzene, e-diphenyldiazene, azobisbenzene, azofume, 1,2-diphenyldiazene, azodibenzeneazofume, diphenyldiimide, azobenzide
CHEBI:190358
C1=CC=C(C=C1)N=NC1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0361

RTECSNumber : CN1400000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions