missing translation for 'onlineSavingsMsg'
Learn More

Azobenzene 98.0+%, TCI America™

Catalog No. p-7116961
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A056525G 25 g
A0565100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A056525G Supplier TCI America Supplier No. A056525G
Only null left

Chemical Identifiers

CAS 103-33-3
Molecular Formula C12H10N2
Molecular Weight (g/mol) 182.23
MDL Number MFCD00003022
InChI Key DMLAVOWQYNRWNQ-UHFFFAOYSA-N
Synonym azobenzene, diazene, diphenyl, diazobenzene, e-diphenyldiazene, azobisbenzene, azofume, 1,2-diphenyldiazene, azodibenzeneazofume, diphenyldiimide, azobenzide
PubChem CID 2272
ChEBI CHEBI:190358
IUPAC Name diphenyldiazene
SMILES C1=CC=C(C=C1)N=NC1=CC=CC=C1

Specifications

Melting Point 68°C
Boiling Point 293°C
Quantity 25 g
UN Number 3077
Formula Weight 182.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Azobenzene
EINECSNumber (3)-0361
RTECSNumber CN1400000
TSCA Yes