missing translation for 'onlineSavingsMsg'
Learn More

Azobenzene-3,3'-dicarboxylic Acid 95.0+%, TCI America™

Catalog No. A15981G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15981G 1 g
A15985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15981G Supplier TCI America Supplier No. A15981G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 621-18-1
Molecular Formula C14H10N2O4
Molecular Weight (g/mol) 270.244
MDL Number MFCD03844759
InChI Key QBVIXYABUQQSRY-UHFFFAOYSA-N
PubChem CID 578067
IUPAC Name 3-[(3-carboxyphenyl)diazenyl]benzoic acid
SMILES C1=CC(=CC(=C1)N=NC2=CC=CC(=C2)C(=O)O)C(=O)O

Specifications

Quantity 1 g
Synonym 3,3′C-Azodibenzoic Acid
Formula Weight 270.24
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Azobenzene-3,3′-dicarboxylic Acid