accessibility menu, dialog, popup

UserName

CAS RN 621-18-1

IUPAC Name: 3-[2-(3-carboxyphenyl)diazen-1-yl]benzoic acid
Synonyms: 3-[2-(3-carboxyphenyl)diazen-1-yl]benzoic acid
Molecular Weight (g/mol): 270.24
Molecular Formula: C14H10N2O4
InChi Key: QBVIXYABUQQSRY-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC=C1)N=NC1=CC=CC(=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results