missing translation for 'onlineSavingsMsg'
Learn More

Dynalon™ Azlon™ Solvent Venting Wash Bottles

Catalog No. p-46182
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Closure Color:
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Color
034059 250 mL White
034058 500 mL White
03406155 1000 mL White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 034059 Supplier Dynalon™ Supplier No. 5064750001
Only null left

Unique two-way valve stop vapour pressure build-up and eliminates the hazard of volatile solvent leaks

Translucent low density polyethylene wash bottles feature unique two-way valve that stops vapor pressure build-up and eliminates the hazard of volatile solvent leaks. Wide neck provides for easier and safer filling. Plus, for quick identification, cadmium-free polypropylene dispensing closures are color coded. And, for optimum control in dispensing, the jet is precision molded. Each bottle features a clear, indelible label describing the contents, international hazard codes and instructional diagrams for safe usage. Unlabeled bottles with write-on label areas and complete set of adhesive hazard labels for custom use are also available. Supplied in convenient shelf packs of five.

Order Info

Bottles are available in 250 or 500mL capacities with five common solvent labels.

Compliance

All bottles have SPI recycling code stamped on base.

Specifications

Capacity 8 oz., 250 mL
Capacity (Metric) 250 mL
Capacity (English) 8 oz.
Color Translucent
Material LDPE
Packaging PK/5
Quantity PK/5
Sterility Non-sterile
Width (Metric) 58.92mm
Closure Color White
Certifications/Compliance SPI recycling code stamped on base
Height (Metric) 206.24 mm
Product Type Wash Bottle
Width (English) 2.32 in.
Height (English) 8.12 in.
Chemical Label Custom Write-on
Closure Material Polypropylene
Autoclavable Not Autoclavable
Show More Show Less