Learn More
Dynalon™ Azlon™ Polypropylene Carboy with Spigot
For Use With: Storing and dispensing liquids in educational settings.; Autoclavable; Closure Material: Polypropylene; Color: White
Supplier:  Dynalon™ 5056343
 View more versions of this product
Catalog No. S01989B
Only null left
Description
Description
  • Autoclavable polypropylene™, heavy-duty construction
  • Stylish rounded octagonal shape for increased rigidity when full
  • Offset neck for easier filling and pouring
  • Integral, comfortable blow molded handles for ease of carrying, are end-sealed to prevent contamination
  • Heavy-duty closures
  • Carboys feature a durable and leakproof heavy-duty spigot with boss fitting for extra strength
  • Spigot features removable tube fitting for added convenience
  • Molded graduations allow easy viewing of contents
Specifications
Specifications
Autoclavable
20 L
White
Blow molded handles
Offset
Rounded Octagonal
5.3 gal.
Polypropylene
Storing and dispensing liquids in educational settings.
Polypropylene
Carboy with Spigot
Product Suggestions
Product Suggestions