missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azithromycin Dihydrate 98.0+%, TCI America™
Supplier: TCI America A20765G
Description
Azithromycin Dihydrate 98.0+%
Specifications
| Azithromycin Dihydrate | |
| White | |
| MFCD01862248 | |
| azithromycin hydrate | |
| CCC1C(C(C(N(CC(CC(C(C(C(C(C(=O)O1)C)OC2CC(C(C(O2)C)O)(C)OC)C)OC3C(C(CC(O3)C)N(C)C)O)(C)O)C)C)C)O)(C)O.O.O | |
| 785.026 | |
| 749.00 | |
| Crystalline Powder |
| 117772-70-0 | |
| C38H76N2O14 | |
| 5 g | |
| SRMPHJKQVUDLQE-GTCGPNQGSA-N | |
| (2R,3S,4S,5R,8R,10R,11R,12S,13R,14R)-11-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-2-ethyl-3,4,10-trihydroxy-13-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-3,5,6,8,10,12,14-heptamethyl-1-oxa-6-azacyclopentadecan-15-o | |
| 131845180 | |
| ≥98.0% (N) |
Chemical Identifiers
| 117772-70-0 | |
| 785.026 | |
| SRMPHJKQVUDLQE-GTCGPNQGSA-N | |
| 131845180 | |
| CCC1C(C(C(N(CC(CC(C(C(C(C(C(=O)O1)C)OC2CC(C(C(O2)C)O)(C)OC)C)OC3C(C(CC(O3)C)N(C)C)O)(C)O)C)C)C)O)(C)O.O.O |
| C38H76N2O14 | |
| MFCD01862248 | |
| azithromycin hydrate | |
| (2R,3S,4S,5R,8R,10R,11R,12S,13R,14R)-11-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-2-ethyl-3,4,10-trihydroxy-13-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-3,5,6,8,10,12,14-heptamethyl-1-oxa-6-azacyclopentadecan-15-o |
Safety and Handling
TSCA : No