missing translation for 'onlineSavingsMsg'
Learn More

Azithromycin Dihydrate 98.0+%, TCI America™

Catalog No. p-7117716
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20761G 1 g
A20765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20761G Supplier TCI America Supplier No. A20761G

Azithromycin Dihydrate 98.0+%

Chemical Identifiers

CAS 117772-70-0
Molecular Formula C38H76N2O14
Molecular Weight (g/mol) 785.026
MDL Number MFCD01862248
InChI Key SRMPHJKQVUDLQE-GTCGPNQGSA-N
Synonym azithromycin hydrate
PubChem CID 131845180
IUPAC Name (2R,3S,4S,5R,8R,10R,11R,12S,13R,14R)-11-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-2-ethyl-3,4,10-trihydroxy-13-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-3,5,6,8,10,12,14-heptamethyl-1-oxa-6-azacyclopentadecan-15-o
SMILES CCC1C(C(C(N(CC(CC(C(C(C(C(C(=O)O1)C)OC2CC(C(C(O2)C)O)(C)OC)C)OC3C(C(CC(O3)C)N(C)C)O)(C)O)C)C)C)O)(C)O.O.O

Specifications

Color White
Quantity 1 g
Formula Weight 749.00
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Azithromycin Dihydrate