missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azilsartan 98.0+%, TCI America™
$214.37 - $643.10
Chemical Identifiers
| CAS | 147403-03-0 |
|---|---|
| Molecular Formula | C25H20N4O5 |
| Molecular Weight (g/mol) | 456.46 |
| MDL Number | MFCD20278186 |
| InChI Key | KGSXMPPBFPAXLY-UHFFFAOYSA-N |
| Synonym | azilsartan, unii-f9nux55p23, azilsartan tak-536, chembl57242, 2-ethoxy-1-2'-5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl-1h-benzimidazole-7-carboxylic acid, 1h-benzimidazole-7-carboxylic acid, 1-2'-2,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl 1,1'-biphenyl-4-yl methyl-2-ethoxy, 1-2'-2,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl 1,1'-biphenyl-4-yl methyl-2-ethoxy-1h-benzimidazole-7-carboxylic acid, 2-ethoxy-1-2'-5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl-1h-benzo d imidazole-7-carboxylic acid, 2-ethoxy-1-2'-4,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl benzimidazole-7-carboxylic acid, 2-ethoxy-3-4-2-5-oxo-2h-1,2,4-oxadiazol-3-yl phenyl phenyl methyl benzimidazole-4-carboxylic acid |
| PubChem CID | 9825285 |
| ChEBI | CHEBI:68850 |
| IUPAC Name | 2-ethoxy-1-{[2'-(5-oxo-2,5-dihydro-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-yl]methyl}-1H-1,3-benzodiazole-7-carboxylic acid |
| SMILES | CCOC1=NC2=CC=CC(C(O)=O)=C2N1CC1=CC=C(C=C1)C1=CC=CC=C1C1=NC(=O)ON1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A2601200MG
|
TCI America
A2601200MG |
200 mg |
Each for $214.37
|
|
|||||
|
A26011G
|
TCI America
A26011G |
1 g |
Each for $643.10
|
|
|||||
Chemical Identifiers
| 147403-03-0 | |
| 456.46 | |
| KGSXMPPBFPAXLY-UHFFFAOYSA-N | |
| 9825285 | |
| 2-ethoxy-1-{[2'-(5-oxo-2,5-dihydro-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-yl]methyl}-1H-1,3-benzodiazole-7-carboxylic acid |
| C25H20N4O5 | |
| MFCD20278186 | |
| azilsartan, unii-f9nux55p23, azilsartan tak-536, chembl57242, 2-ethoxy-1-2'-5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl-1h-benzimidazole-7-carboxylic acid, 1h-benzimidazole-7-carboxylic acid, 1-2'-2,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl 1,1'-biphenyl-4-yl methyl-2-ethoxy, 1-2'-2,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl 1,1'-biphenyl-4-yl methyl-2-ethoxy-1h-benzimidazole-7-carboxylic acid, 2-ethoxy-1-2'-5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl-1h-benzo d imidazole-7-carboxylic acid, 2-ethoxy-1-2'-4,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl benzimidazole-7-carboxylic acid, 2-ethoxy-3-4-2-5-oxo-2h-1,2,4-oxadiazol-3-yl phenyl phenyl methyl benzimidazole-4-carboxylic acid | |
| CHEBI:68850 | |
| CCOC1=NC2=CC=CC(C(O)=O)=C2N1CC1=CC=C(C=C1)C1=CC=CC=C1C1=NC(=O)ON1 |
Specifications
| 147403-03-0 | |
| White | |
| MFCD20278186 | |
| azilsartan, unii-f9nux55p23, azilsartan tak-536, chembl57242, 2-ethoxy-1-2'-5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl-1h-benzimidazole-7-carboxylic acid, 1h-benzimidazole-7-carboxylic acid, 1-2'-2,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl 1,1'-biphenyl-4-yl methyl-2-ethoxy, 1-2'-2,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl 1,1'-biphenyl-4-yl methyl-2-ethoxy-1h-benzimidazole-7-carboxylic acid, 2-ethoxy-1-2'-5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl-1h-benzo d imidazole-7-carboxylic acid, 2-ethoxy-1-2'-4,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl benzimidazole-7-carboxylic acid, 2-ethoxy-3-4-2-5-oxo-2h-1,2,4-oxadiazol-3-yl phenyl phenyl methyl benzimidazole-4-carboxylic acid | |
| CCOC1=NC2=CC=CC(C(O)=O)=C2N1CC1=CC=C(C=C1)C1=CC=CC=C1C1=NC(=O)ON1 | |
| 456.46 | |
| CHEBI:68850 | |
| ≥98.0% (HPLC,T) | |
| Azilsartan |
| 188°C | |
| C25H20N4O5 | |
| 200 mg | |
| KGSXMPPBFPAXLY-UHFFFAOYSA-N | |
| 2-ethoxy-1-{[2'-(5-oxo-2,5-dihydro-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-yl]methyl}-1H-1,3-benzodiazole-7-carboxylic acid | |
| 9825285 | |
| 456.46 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator