missing translation for 'onlineSavingsMsg'
Learn More

Azilsartan 98.0+%, TCI America™

Supplier:  TCI America A2601200MG

 View more versions of this product

Catalog No. A2601200MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Azilsartan
188°C
C25H20N4O5
200 mg
KGSXMPPBFPAXLY-UHFFFAOYSA-N
2-ethoxy-1-{[2'-(5-oxo-2,5-dihydro-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-yl]methyl}-1H-1,3-benzodiazole-7-carboxylic acid
9825285
456.46
Crystalline Powder
147403-03-0
White
MFCD20278186
azilsartan, unii-f9nux55p23, azilsartan tak-536, chembl57242, 2-ethoxy-1-2'-5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl-1h-benzimidazole-7-carboxylic acid, 1h-benzimidazole-7-carboxylic acid, 1-2'-2,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl 1,1'-biphenyl-4-yl methyl-2-ethoxy, 1-2'-2,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl 1,1'-biphenyl-4-yl methyl-2-ethoxy-1h-benzimidazole-7-carboxylic acid, 2-ethoxy-1-2'-5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl-1h-benzo d imidazole-7-carboxylic acid, 2-ethoxy-1-2'-4,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl benzimidazole-7-carboxylic acid, 2-ethoxy-3-4-2-5-oxo-2h-1,2,4-oxadiazol-3-yl phenyl phenyl methyl benzimidazole-4-carboxylic acid
CCOC1=NC2=CC=CC(C(O)=O)=C2N1CC1=CC=C(C=C1)C1=CC=CC=C1C1=NC(=O)ON1
456.46
CHEBI:68850
≥98.0% (HPLC,T)

Chemical Identifiers

147403-03-0
456.46
KGSXMPPBFPAXLY-UHFFFAOYSA-N
9825285
2-ethoxy-1-{[2'-(5-oxo-2,5-dihydro-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-yl]methyl}-1H-1,3-benzodiazole-7-carboxylic acid
C25H20N4O5
MFCD20278186
azilsartan, unii-f9nux55p23, azilsartan tak-536, chembl57242, 2-ethoxy-1-2'-5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl-1h-benzimidazole-7-carboxylic acid, 1h-benzimidazole-7-carboxylic acid, 1-2'-2,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl 1,1'-biphenyl-4-yl methyl-2-ethoxy, 1-2'-2,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl 1,1'-biphenyl-4-yl methyl-2-ethoxy-1h-benzimidazole-7-carboxylic acid, 2-ethoxy-1-2'-5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl-1h-benzo d imidazole-7-carboxylic acid, 2-ethoxy-1-2'-4,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl benzimidazole-7-carboxylic acid, 2-ethoxy-3-4-2-5-oxo-2h-1,2,4-oxadiazol-3-yl phenyl phenyl methyl benzimidazole-4-carboxylic acid
CHEBI:68850
CCOC1=NC2=CC=CC(C(O)=O)=C2N1CC1=CC=C(C=C1)C1=CC=CC=C1C1=NC(=O)ON1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions