missing translation for 'onlineSavingsMsg'
Learn More

Azidoacetic Acid 97.0+%, TCI America™

Catalog No. a30791g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A30791G Supplier TCI America Supplier No. A30791G
Only null left

Chemical Identifiers

CAS 18523-48-3
Molecular Formula C2H3N3O2
Molecular Weight (g/mol) 101.065
MDL Number MFCD01320907
InChI Key PPXUUPXQWDQNGO-UHFFFAOYSA-N
PubChem CID 155605
IUPAC Name 2-azidoacetic acid
SMILES C(C(=O)O)N=[N+]=[N-]

Specifications

Melting Point 16°C
Boiling Point 116°C
Quantity 1 g
Formula Weight 101.07
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Azidoacetic Acid
TSCA No
Recommended Storage Refrigerator