accessibility menu, dialog, popup

UserName

CAS RN 18523-48-3

IUPAC Name: 2-azidoacetic acid
Synonyms: azidoacetic acid
Molecular Weight (g/mol): 101.07
Molecular Formula: C2H3N3O2
InChi Key: PPXUUPXQWDQNGO-UHFFFAOYSA-N
SMILES: OC(=O)CN=[N+]=[N-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results