missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azido-PEG4-Amine 95.0+%, TCI America™
Supplier: TCI America A3004250MG
Specifications
| Azido-PEG4-Amine | |
| Yellow | |
| MFCD20134134 | |
| 2735 | |
| ZMBGKXBIVYXREN-UHFFFAOYSA-N | |
| 14-azido-3,6,9,12-tetraoxatetradecan-1-amine | |
| 71313220 | |
| ≥95.0% (GC) |
| 951671-92-4 | |
| C10H22N4O4 | |
| 250 mg | |
| Amino-PEG4-Azide, 14-Azido-3,6,9,12-tetraoxatetradecan-1-amine, 14-Amino-3,6,9,12-tetraoxatetradecanyl Azide, 1-Amino-14-azido-3,6,9,12-tetraoxatetradecane | |
| NCCOCCOCCOCCOCCN=[N+]=[N-] | |
| 262.31 | |
| 262.31 | |
| Liquid |
Chemical Identifiers
| 951671-92-4 | |
| 262.31 | |
| ZMBGKXBIVYXREN-UHFFFAOYSA-N | |
| 71313220 | |
| NCCOCCOCCOCCOCCN=[N+]=[N-] |
| C10H22N4O4 | |
| MFCD20134134 | |
| Amino-PEG4-Azide, 14-Azido-3,6,9,12-tetraoxatetradecan-1-amine, 14-Amino-3,6,9,12-tetraoxatetradecanyl Azide, 1-Amino-14-azido-3,6,9,12-tetraoxatetradecane | |
| 14-azido-3,6,9,12-tetraoxatetradecan-1-amine |
Safety and Handling
TSCA : No
Recommended Storage : Freezer (-20 °C)