accessibility menu, dialog, popup

UserName

CAS RN 951671-92-4

IUPAC Name: 14-azido-3,6,9,12-tetraoxatetradecan-1-amine
Synonyms: 14-azido-3,6,9,12-tetraoxatetradecan-1-amine
Molecular Weight (g/mol): 262.31
Molecular Formula: C10H22N4O4
InChi Key: ZMBGKXBIVYXREN-UHFFFAOYSA-N
SMILES: NCCOCCOCCOCCOCCN=[N+]=[N-]
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results