missing translation for 'onlineSavingsMsg'
Learn More

Azido-PEG4-Amine 95.0+%, TCI America™

Catalog No. A300450MG
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A300450MG 50 mg
A3004250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A300450MG Supplier TCI America Supplier No. A300450MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 951671-92-4
Molecular Formula C10H22N4O4
Molecular Weight (g/mol) 262.31
MDL Number MFCD20134134
InChI Key ZMBGKXBIVYXREN-UHFFFAOYSA-N
Synonym Amino-PEG4-Azide, 14-Azido-3,6,9,12-tetraoxatetradecan-1-amine, 14-Amino-3,6,9,12-tetraoxatetradecanyl Azide, 1-Amino-14-azido-3,6,9,12-tetraoxatetradecane
PubChem CID 71313220
IUPAC Name 14-azido-3,6,9,12-tetraoxatetradecan-1-amine
SMILES NCCOCCOCCOCCOCCN=[N+]=[N-]

Specifications

Color Yellow
Quantity 50 mg
UN Number 2735
Formula Weight 262.31
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Azido-PEG4-Amine
TSCA No
Recommended Storage Freezer (-20 °C)