missing translation for 'onlineSavingsMsg'
Learn More

Azido-PEG3-carboxyrhodamine 110 conjugate, Thermo Scientific Chemicals

Catalog No. p-7043021
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65107LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ65107LB0 Supplier Thermo Scientific Chemicals Supplier No. J65107LB0
Only null left

It is suitable for confocal laser scanning microscopy. Its long PEG spacer should reduce any steric effect of the dye on the molecule to be labeled. Carboxyrhodamine 110 and its derivatives are superior alternatives to other green fluorescent dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is suitable for confocal laser scanning microscopy. Its long PEG spacer should reduce any steric effect of the dye on the molecule to be labeled. Carboxyrhodamine 110 and its derivatives are superior alternatives to other green fluorescent dyes.

Solubility
Soluble in DMSO, DMF or methanol.

Notes
Excite using the 488nm spectral line of the argon ion laser

Chemical Identifiers

CAS 1536327-95-3
Molecular Formula C29H30N6O7
MDL Number MFCD22380746
Synonym Azide-Fluor 488

Specifications

Color Red
Quantity 5 mg
Solubility Information Soluble in DMSO,DMF or methanol.
Formula Weight 574.59
Physical Form Solid
Chemical Name or Material Azido-PEG3-carboxyrhodamine 110 conjugate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only