Learn More
Azetidine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 296730050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Azetidine | |
| 0.8500g/mL | |
| −20°C | |
| 97.5% min. (GC) | |
| C3H7N | |
| 5g | |
| 0.85 | |
| Solubility in water: miscible. | |
| C1CNC1 | |
| 57.09 | |
| CHEBI:30968 | |
| 98% |
| 503-29-7 | |
| 61.0°C to 62.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4290 to 1.4330 | |
| 20,2 | |
| azacyclobutane, trimethylene imine, 1,3-propylenimine, trimethylenimine, azetidin, unii-37s883xdwr, azetidine, l, polypropylene amine, azete, tetrahydro, azetidine | |
| HONIICLYMWZJFZ-UHFFFAOYSA-N | |
| azetidine | |
| 10422 | |
| 57.09 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapour.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 207-963-8