accessibility menu, dialog, popup

UserName

CAS RN 503-29-7

IUPAC Name: azetidine
Synonyms: azetidine
Molecular Weight (g/mol): 57.1
Molecular Formula: C3H7N
InChi Key: HONIICLYMWZJFZ-UHFFFAOYSA-N
SMILES: C1CNC1
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)