missing translation for 'onlineSavingsMsg'
Learn More

Azer Scientific Pre-Filled 10% Neutral Buffered Formalin

A traditional fixative buffered to a neutral pH

Supplier:  Azer Scientific PFNBF240

 View more versions of this product

Catalog No. 22026446


Only null left

Description

Description

  • Designed specifically for the collection and transport of various sizes of tissue samples
  • Each container is half filled with 10% neutral buffered formalin
  • Packaged with our proprietary Sure-Lok™ system

Histological applications for fixing and storing tissue specimens

Compliance

These containers are 95 kPa compliant

Specifications

Specifications

8.1 oz., pre-filed 4.1 oz.
240 mL, pre-filled 120 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions