missing translation for 'onlineSavingsMsg'
Learn More

Azer Scientific Pre-Filled 10% Neutral Buffered Formalin

Catalog No. p-3796420
Change view
Click to view available options
Capacity (Metric):
1000 mL, pre-filled 500 mL
180 mL, pre-filled 90 mL
240 mL, pre-filled 120 mL
480 mL, pre-filled 240 mL
60 mL, pre-filled 30 mL
Capacity (English):
16.2 oz., pre-filled 8.1 oz.
2 oz., pre-filled 50%
33.8 oz., pre-filled 16.9 oz.
6 oz., pre-filled 50%
8.1 oz., pre-filed 4.1 oz.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Capacity (English)
22026445 180 mL, pre-filled 90 mL 6 oz., pre-filled 50%
22026439 60 mL, pre-filled 30 mL 2 oz., pre-filled 50%
22026446 240 mL, pre-filled 120 mL 8.1 oz., pre-filed 4.1 oz.
22026448 480 mL, pre-filled 240 mL 16.2 oz., pre-filled 8.1 oz.
22026449 1000 mL, pre-filled 500 mL 33.8 oz., pre-filled 16.9 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22026445 Supplier Azer Scientific Supplier No. PFNBF180
Only null left

A traditional fixative buffered to a neutral pH

  • Designed specifically for the collection and transport of various sizes of tissue samples
  • Each container is half filled with 10% neutral buffered formalin
  • Packaged with our proprietary Sure-Lok™ system

Histological applications for fixing and storing tissue specimens

Compliance

These containers are 95 kPa compliant

Specifications

Capacity (English) 6 oz., pre-filled 50%
Capacity (Metric) 180 mL, pre-filled 90 mL