missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azelnidipine 98.0+%, TCI America™
$145.40 - $501.43
Chemical Identifiers
| CAS | 123524-52-7 |
|---|---|
| Molecular Formula | C33H34N4O6 |
| Molecular Weight (g/mol) | 582.66 |
| MDL Number | MFCD00865803 |
| InChI Key | ZKFQEACEUNWPMT-UHFFFAOYNA-N |
| Synonym | 2-Amino-1,4-dihydro-6-methyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 3-[1-(Diphenylmethyl)-3-azetidinyl] 5-Isopropyl Ester, 3-[1-(Diphenylmethyl)-3-azetidinyl] 5-Isopropyl 2-Amino-1,4-dihydro-6-methyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate |
| PubChem CID | 65948 |
| IUPAC Name | 3-[1-(diphenylmethyl)azetidin-3-yl] 5-propan-2-yl 2-amino-6-methyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate |
| SMILES | CC(C)OC(=O)C1=C(C)NC(N)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC1CN(C1)C(C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 123524-52-7 | |
| 582.66 | |
| ZKFQEACEUNWPMT-UHFFFAOYNA-N | |
| 65948 | |
| CC(C)OC(=O)C1=C(C)NC(N)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC1CN(C1)C(C1=CC=CC=C1)C1=CC=CC=C1 |
| C33H34N4O6 | |
| MFCD00865803 | |
| 2-Amino-1,4-dihydro-6-methyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 3-[1-(Diphenylmethyl)-3-azetidinyl] 5-Isopropyl Ester, 3-[1-(Diphenylmethyl)-3-azetidinyl] 5-Isopropyl 2-Amino-1,4-dihydro-6-methyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate | |
| 3-[1-(diphenylmethyl)azetidin-3-yl] 5-propan-2-yl 2-amino-6-methyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate |
Specifications
| 123524-52-7 | |
| C33H34N4O6 | |
| 1 g | |
| ZKFQEACEUNWPMT-UHFFFAOYNA-N | |
| 3-[1-(diphenylmethyl)azetidin-3-yl] 5-propan-2-yl 2-amino-6-methyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate | |
| 65948 | |
| ≥98.0% (HPLC,T) | |
| Azelnidipine |
| Yellow | |
| MFCD00865803 | |
| 2-Amino-1,4-dihydro-6-methyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 3-[1-(Diphenylmethyl)-3-azetidinyl] 5-Isopropyl Ester, 3-[1-(Diphenylmethyl)-3-azetidinyl] 5-Isopropyl 2-Amino-1,4-dihydro-6-methyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate | |
| CC(C)OC(=O)C1=C(C)NC(N)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC1CN(C1)C(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 582.66 | |
| 582.66 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : US7968332
TSCA : No